Molybdenum standard dissolved in oil, 1.000 µg/g Mo in mineral oil Specpure® til wear metal analysis

Leverandør: Thermo Fisher Scientific

14213.18
14213.18EA 1100 NOK
14213.18
Molybdenum standard dissolved in oil, 1.000 µg/g Mo in mineral oil Specpure® til wear metal analysis
Standarder Kationstandarder Molybdenum Standards

Physical Form: Liquid
MDL No.: MFCD00003465

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR