Magnesium standard dissolved in oil, 1.000 µg/g Mg in mineral oil Specpure® til wear metal analysis

Leverandør: Thermo Fisher Scientific

14219.1Y 14219.18
14219.1YEA 0 NOK
14219.1Y 14219.18
Magnesium standard dissolved in oil, 1.000 µg/g Mg in mineral oil Specpure® til wear metal analysis
Standarder Kationstandarder Magnesium Standards

Physical Form: Liquid
MDL No.: MFCD00085308

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00085308

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR