Iron standard dissolved in oil, 1.000 µg/g Fe in mineral oil Specpure® til wear metal analysis

Leverandør: Thermo Fisher Scientific

14220.18
14220.18EA 1475 NOK
14220.18
Iron standard dissolved in oil, 1.000 µg/g Fe in mineral oil Specpure® til wear metal analysis
Standarder Kationstandarder Iron Standards

Physical form: liquid
MDL No.: MFCD00010999

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR