Bismuth standard dissolved in oil, 1.000 µg/g Bi in mineral oil Specpure® til wear metal analysis

Leverandør: Thermo Fisher Scientific

14223.18
14223.18EA 1700 NOK
14223.18
Bismuth standard dissolved in oil, 1.000 µg/g Bi in mineral oil Specpure® til wear metal analysis
Standarder Kationstandarder Bismuth Standards

Physical form: liquid
MDL No.: MFCD00134033

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR