Acetonitril med 0,1% (v/v) Maursyre ( metansyre), CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™

Leverandør: Honeywell Chemicals
Danger

14272-2L
14272-2LEA 6550 NOK
14272-2L
Acetonitril med 0,1% (v/v) Maursyre ( metansyre), CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Acetonitril med 0,1% Maursyre ( metansyre)

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise

Kokepunkt: 81 °C
Smeltepunkt: –46 °C
Tetthet: 0,78 g/cm³ (20 °C)
Flash punkt: 5 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001878
EINECS: 200-835-2
FN: 1648
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Formic acid 0.095 - 0.105 Vol.%
Non-volatile matter Max. 0.0001 %
Water (Karl Fischer) Max. 0.02 %
Aluminium (Al) Max. 0.03 ppm
Calcium (Ca) Max. 0.1 ppm
Iron (Fe) Max. 0.05 ppm
Potassium (K) Max. 0.1 ppm
Magnesium (Mg) Max. 0.03 ppm
Sodium (Na) Max. 0.1 ppm
Transmittance at 254 nm Min. 90 %
Transmittance at 230 nm Min. 15 %
Transmittance at 210 nm Min. 5 %
Suitability for the UHPLC-MS complying
UHPLC Gradient drift 254 nm Max. 30 mAU
UHPLC Gradient peak 254 nm Max. 2 mAU
UHPLC-MS ESI(+) (Reserpine) Max. 20 ppb
Fluorescence (chinin) at 365 nm Max. 0.5 ppb
Fluorescence (chinin) at 254 nm Max. 0.5 ppb

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR