Methanol with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™

Leverandør: Honeywell Chemicals
Danger

14276-2L
14276-2LEA 1875 NOK
14276-2L
Methanol with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Methanol with 0,1% (v/v) formic acid

Solvent blend contains 0,095 to 0,105% (v/v) formic acid (T), tested for UHPLC-MS.


  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise

Smeltepunkt: −46 °C
Flash punkt: 11 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004595
EINECS: 200-659-6
FN: 1230
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Formic acid 0.095 - 0.105 Vol.%
Non-volatile matter Max. 0.0001 %
Calcium (Ca) Max. 0.3 ppm
Potassium (K) Max. 0.3 ppm
Magnesium (Mg) Max. 0.3 ppm
Sodium (Na) Max. 1 ppm
Transmittance at 254 nm Min. 90 %
Transmittance at 230 nm Min. 10 %
Suitability for the UHPLC-MS complying
UHPLC Gradient drift 254 nm Max. 30 mAU
UHPLC Gradient peak 254 nm Max. 4 mAU
UHPLC-MS ESI(+) (Reserpine) Max. 20 ppb
Fluorescence (chinin) at 254 nm Max. 0.7 ppb
Fluorescence (chinin) at 365 nm Max. 0.5 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR