Boron 10000 mg/L B in ammonium hydroxide solution (max. 1%) (fra B(OH₃)) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

14370.AD 14370.AE 14370.AP
14370.ADEA 1325 NOK
14370.AD 14370.AE 14370.AP
Boron 10000 mg/L B in ammonium hydroxide solution (max. 1%) (fra B(OH₃)) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Boron Standards

Molecular Formula: H₃BO₃ in 1% NH₄OH
Physical Form: Liquid
MDL No.: MFCD00134034


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR