Cerium10000 mg/L Ce in 5% salpetersyre (fra Ce) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

14371.AD 14371.AE 14371.AP
14371.ADEA 1325 NOK
14371.AD 14371.AE 14371.AP
Cerium10000 mg/L Ce in 5% salpetersyre (fra Ce) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Cerium Standards

Molecular Formula: Ce in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010924


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR