Terbium 10000 mg/L Tb in 5% salpetersyre (fra Tb₄O₇) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

14383.AD 14383.AE 14383.AP
14383.ADEA 910 NOK
14383.AD 14383.AE 14383.AP
Terbium 10000 mg/L Tb in 5% salpetersyre (fra Tb₄O₇) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Terbium Standards

Molecular formula: Tb₄O₇ in 5% HNO₃
Physical form: liquid
UN No.: 3264
MDL No.: MFCD00011256


Traceable to NIST


Sertifiseringer: Traceable to NIST.

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00011256

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR