Kobalt 10000 mg/L Co in 5% salpetersyre (fra Co) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

14390.AD 14390.AE 14390.AP
14390.ADEA 1450 NOK
14390.AD 14390.AE 14390.AP
Kobalt 10000 mg/L Co in 5% salpetersyre (fra Co) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Cobalt Standards

Molecular Formula: Co in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010935


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR