Kalsium 10000 mg/L Ca in 5% salpetersyre (fra CaCO₃) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

14407.AD 14407.AE 14407.AP
14407.ADEA 1550 NOK
14407.AD 14407.AE 14407.AP
Kalsium 10000 mg/L Ca in 5% salpetersyre (fra CaCO₃) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Calcium Standards

Molecular Formula: CaCO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085314


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR