Germanium 10000 mg/L Ge in 5% salpetersyre with hydrofluoric acid (max. 1%) (de Ge) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

14410.AD 14410.AE 14410.AP
14410.ADEA 1600 NOK
14410.AD 14410.AE 14410.AP
Germanium 10000 mg/L Ge in 5% salpetersyre with hydrofluoric acid (max. 1%) (de Ge) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Germanium Standards

Molecular Formula: Ge in 5% HNO₃/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085310


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR