Tantalum 10000 mg/L Ta in 2% hydrofluoric acid (fra TaCl₅) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

14419.AD 14419.AE 14419.AP
14419.ADEA 2075 NOK
14419.AD 14419.AE 14419.AP
Tantalum 10000 mg/L Ta in 2% hydrofluoric acid (fra TaCl₅) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Tantalum Standards

Molecular Formula: TaCl₅ in 2% HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011252


Sertifiseringer: Traceable to NIST

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00011252

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR