Yttrium 10000 mg/L Y in 5% salpetersyre (fra Y₂O₃) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

14422.AD 14422.AE 14422.AP
14422.ADEA 1425 NOK
14422.AD 14422.AE 14422.AP
Yttrium 10000 mg/L Y in 5% salpetersyre (fra Y₂O₃) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Yttrium Standards

Molecular Formula: Y₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011468


Sertifiseringer: Traceable to NIST

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00011468

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR