Gallium 10000 mg/L Ga in 5% salpetersyre with saltsyre (max. 1%) (fra Ga) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

14427.AD 14427.AE 14427.AP
14427.ADEA 1625 NOK
14427.AD 14427.AE 14427.AP
Gallium 10000 mg/L Ga in 5% salpetersyre with saltsyre (max. 1%) (fra Ga) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Gallium Standards

Molecular Formula: Ga in 5% HNO₃/tr. HCl
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134045


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR