Lantan 10000 mg/L La in 5% salpetersyre (fra La₂O₃) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

14429.AP 14429.AD 14429.AE
14429.APEA 5050 NOK
14429.AP 14429.AD 14429.AE
Lantan 10000 mg/L La in 5% salpetersyre (fra La₂O₃) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Lanthanum Standards

Molecular Formula: La₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011066


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR