Neodymium 10000 mg/L Nd in 5% salpetersyre (fra Nd₂O₃) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

14431.AE 14431.AD 14431.AP
14431.AEEA 2600 NOK
14431.AE 14431.AD 14431.AP
Neodymium 10000 mg/L Nd in 5% salpetersyre (fra Nd₂O₃) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Neodymium Standards

Molecular Formula: Nd₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011130


Sertifiseringer: Traceable to NIST

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00011130

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR