Svovel 10000 mg/L i vann (fra (NH₄)₂SO₄) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

14436.AD 14436.AE 14436.AP
14436.ADEA 1450 NOK
14436.AD 14436.AE 14436.AP
Svovel 10000 mg/L i vann (fra (NH₄)₂SO₄) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Sulphur Standards

Molecular Formula: (NH₄)₂SO₄ in H₂O
Physical Form: Liquid
MDL No.: MFCD00085316


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR