Ytterbium triacetate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Leverandør: Thermo Fisher Scientific

14576.09 14576.18
14576.09EA 1050 NOK
14576.09 14576.18
Ytterbium triacetate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Ytterbium triacetate hydrate

Purity: 99.9% CAS No.: 15280-58-7 Molecular Formula: Yb(OOCCH3)3.xH2O Formula Weight: 350.16(anhy) Physical Form: Crystalline Aggregates Odour: Odourless MDL No.: MFCD00150127

Formel: C₆H₉O₆Yb·H₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150127
CAS-nummer: 15280-58-7
EINECS: 244-137-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR