Graphite electrodes, crater
Leverandør: Thermo Fisher Scientific
14743.KN 40788.KN
14743.KNEA 3625 NOK
14743.KN 40788.KN
Graphite electrodes, crater
Elektroder Elektroder, øvrige
Graphite electrodes are used mainly in electric arc furnace steel production.
  • AGKSP grade
  • Ultra F purity
  • Insoluble in water
They are also used to refine steel in ladle furnaces and in other smelting processes, furnance design, sharp requirements, water spray rings and charging practice.