Sporicide, Minncare® Cold Sterilant
Leverandør: MARCOR PURIFICATION
176-01-125
148-0062EA 27500 NOK
148-0062
Sporicide, Minncare® Cold Sterilant
Rengjøringsløsninger Desinfeksjon
Minncare® Cold Sterilant concentrate is an effective viricide, bactericide, fungicide, and sporicide for surface disinfection in pharmaceutical cleanroom environments. It is completely and contains only pharmaceutical grade raw materials.
  • Leaves no measurable air residue following ventilation of room
  • Faster, safer, and less corrosive than aldehyde-based liquids
  • EPA registered for fogging as part of normal cleaning and disinfecting procedures