Liquid detergent, L900, Nalgene®
Leverandør: Thermo Fisher Scientific
900-4000
148-0114EA 1250 NOK
148-0114
Liquid detergent, L900, Nalgene®
Rengjøringsløsninger Detergenter og overflatevasking
Specifically formulated to clean plastic and glass labware used in pharmaceutical, medical research, academic, biotechnology and other facilities.
  • Formulated to be compatible with all plastics, especially polycarbonate
  • Will not cause crazing, stress cracking or clouding when used as directed
  • For hand or machine use, it is non toxic and low foaming
Non inhibitory according to APHA Standard Methods for the Examination of Water and Wastewater.