Detergents, neodisher® EM, liquid detergents for special diswasher
Leverandør: Dr. Weigert
430033
148-0165EA 2875 NOK
148-0165
Detergents, neodisher® EM, liquid detergents for special diswasher
Rengjøringsløsninger Detergenter og overflatevasking
Neodisher® EM is an additive for emulsification or dispersion as well as foam suppression in special dishwashers in the cosmetics industry, research, pharmaceutical industry and chemical industry.
  • Combination with alkaline and acid cleaners, emulsifies or disperses undissolved impurities
  • Emulsifies and/or disperses fats, oils, paraffins and solid matter in aqueous solutions
  • It also has an anti foaming action