perform® classic concentrate Mucasol, liquid concentrate, universal detergent
Leverandør: Brand
44103
148-0169EA 10125 NOK
148-0169
perform® classic concentrate Mucasol, liquid concentrate, universal detergent
Rengjøringsløsninger Detergenter og overflatevasking
Liquid, alkaline high-performance concentrate for immersion and ultrasonic baths. Universal cleaner for laboratory equipment and instruments made of glass, porcelain, plastic, rubber and metal.
  • Items are cleaned in ~ 10 – 30 minutes, without mechanical help
perform® classic concentrate mucasol replaces chromosulfuric acid, is free from chlorine and caustic alkalis, is non-toxic, nonaggressive and biodegradable. Prolonged exposure of zinc and non-anodized aluminium surfaces should be avoided