Vaskemiddel, pulver for automatisert rengjøring av glassutstyr, neodisher® LaboClean GK
Leverandør: Dr. Weigert
410376
148-3122EA 3025 NOK
148-3122
Vaskemiddel, pulver for automatisert rengjøring av glassutstyr, neodisher® LaboClean GK
Rengjøringsløsninger Detergenter og overflatevasking
neodisher® LaboClean GK is especially suitable for the automated cleaning of glassware for cell and tissue cultures, in the field of microbiology as well as nuclear medicine and isotope laboratories.
  • Mildly alkaline powder especially suitable for removal of residues of culture media, tissues, blood, proteins and organic dyes, free from surfactants
  • Components: <5% chlorinated bleaching agents (organic chlorine carriers); >30% phosphates; 1 to 5% dichloroisocyanuric acid sodium salt
  • Dosage: 2 to 5 g/L
Note: Most of theMore Product Information