Oppvaskmaskin pulver, neodisher® MA
Leverandør: Dr. Weigert
410776
148-3132EA 3975 NOK
148-3132
Oppvaskmaskin pulver, neodisher® MA
Rengjøringsløsninger Detergenter og overflatevasking
Mildly alkaline detergent for washing machines with applications including cleaning of surgical instruments, anaesthesia materials, operating theatre clogs and glassware used in medical, biological and chemical laboratories.
  • Free from oxidising materials
  • Very good cleaning action and very gentle on material
  • Mildly alkaline formulation with defoaming properties
  • Phosphates>30%
  • Non-ionic surfactants<5%
  • Dosage: Typically 1 to 4 g/litre