Vaskemiddel, pulver for automatisert rengjøring av glassutstyr, neodisher® LaboClean UW
Leverandør: Dr. Weigert
410976
148-3183EA 2825 NOK
148-3183
Vaskemiddel, pulver for automatisert rengjøring av glassutstyr, neodisher® LaboClean UW
Rengjøringsløsninger Detergenter og overflatevasking
neodisher® LaboClean UW is suitable for the automated cleaning of laboratory glassware and utensils in water analysis laboratories, in serological institutes and in the phosphates industry. Especially for laboratory glassware used for sensitive analyses, where residues of phosphorous compounds would adulterate analysis results.
  • Mildly alkaline powder for machine washing laboratory glassware
  • Phosphate-free
  • Components: <5% non-ionic surfactants, 5 to 15% oxygenated bleaching agents (perborate)
  • Dosage: 3 to 5 g/L
NB: Most of theMore Product Information