Multi gas detector, GasAlertMax XT II

Leverandør: BW TECHNOLOGIES
XT-0W0M-Y-EU XT-X000-Y-EU XT-XW00-Y-EU XT-XW0M-Y-EU XT-XWHM-Y-EU XT-0W00-Y-EU XT-X00M-Y-EU XT-0W00-Y-UK XT-0W0M-Y-UK XT-X000-Y-UK XT-X00M-Y-UK XT-XW00-Y-UK XT-XW0M-Y-UK XT-XWHM-Y-UK
BWTEXT-0W0M-Y-EUEA 14750 NOK
BWTEXT-0W0M-Y-EU BWTEXT-X000-Y-EU BWTEXT-XW00-Y-EU BWTEXT-XW0M-Y-EU 151-0239 BWTEXT-0W00-Y-EU BWTEXT-X00M-Y-EU BWTEXT-0W00-Y-UK BWTEXT-0W0M-Y-UK BWTEXT-X000-Y-UK BWTEXT-X00M-Y-UK BWTEXT-XW00-Y-UK BWTEXT-XW0M-Y-UK BWTEXT-XWHM-Y-UK
Multi gas detector, GasAlertMax XT II
Gassdeteksjonssystemer
The rugged GasAlert Max XT II reliably monitors up to four gas hazards and combines straightforward one-button field operation with an integrated sampling pump.

  • Compatible with BW MicroDock II automatic test and calibration station
  • Powered by a rechargeable battery pack
  • GasAlert Max XT II comes with standard event logging and datalogging (accessory connectivity kit required)

Tamper-proof, user-adjustable options enable the instrument to be customised to suit your application.

Leveringsinformasjon: Supplied with detector complete with sensors,More Product Information
Bestill nå

SPESIFIKASJONER

Modell GasAlert Max XT II
Environmentally Preferable
Alarm Visual, vibrating, audible (95 dB)
levetid batteri 10 hours at −20 °C, 13 hours at 0 °C
B×D×H 7.0×5.2×13.1 cm
Vekt 316 g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR