Portable gas detectors, GasAlert Micro 5
Leverandør: BW TECHNOLOGIES
M5IRXWBYRDDYE0
151-0248EA 0 NOK
151-0248
Portable gas detectors, GasAlert Micro 5
Gassdeteksjonssystemer
Continuously monitoring up to five gases, the GasAlert Micro 5 was designed and engineered for an extensive range of applications.
  • LCD shows simultaneous gas concentrations
  • Integral motorized pump option for remote sampling
  • Two power options: Alkaline batteries or rechargeable pack
  • Multi-language support in English, French, German, Spanish, and Portuguese
Sertifiseringer: EMI/RFI: EMC 89/336/EEC; IP 65/66; ATEX: X g II 1 G Ga Ex ia IIC T4* X g II 2 G (IR model only); Ex d ia IIC T4* - IECEx: Ga Ex ia IIC T4* Ex d ia IIC T4* (IR model only)
*More Product Information