Multi-gas detector, BW Clip
Leverandør: BW TECHNOLOGIES
BWC4-Y-E BWC4-B-E
151-0316EA 10750 NOK
151-0316 151-0317
Multi-gas detector, BW Clip
Gassdeteksjonssystemer
The BW Clip4 is an easy to clip on four-gas detector (H₂S, CO, O₂, LEL) that is always on, it requires no charging, no battery or sensor replacements.
  • 2-year continuous runtime
  • Durable, ergonomic and wearable
  • Simple one-button operation
  • Intuitive LCD icons
  • Real-time gas concentrations
  • Compatible with IntelliDoX docking system and instrument management system
  • Tested to last in extreme environments
The BW Clip4 is equipped with flashing red non-compliance indicators which warn workers that the detector has not been bump tested according to schedule. ItMore Product Information