Phenoxyethanol, Reference Standard

Leverandør: USP

1526200
1526200.EA 4850 NOK
1526200.
Phenoxyethanol, Reference Standard
Standarder Organiske standarder

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.


Sertifiseringer: US Pharmacopea Reference Standard

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d27cc4d39b806528798164961c23a1285d41d6a9&vid=a46b721cc69101d0c50dc49d9687445b395a3766&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Beskrivelse Phenoxyethanol
Environmentally Preferable
Synonym(s) 2-Phenoxyethanol
CAS-nr 122-99-6

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR