Trifluoreddiksyre 100%, HiPerSolv CHROMANORM® for HPLC
Leverandør: VWR Chemicals
Danger
Synonymer: TFA
Packaged under nitrogen.
153112EEA 2425 NOK
153112E
Trifluoreddiksyre 100%, HiPerSolv CHROMANORM® for HPLC
Trifluoreddiksyre
Formel: C₂HF₃O₂
MW: 114.02 g/mol
Kokepunkt: 71.8 °C (1013 hPa)
Smeltepunkt: –15 °C
Tetthet: 1.490 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004169
CAS-nummer: 76-05-1
EINECS: 200-929-3
FN: 2699
ADR: 8 I
Merck Indeks: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Assay Min. 99.80 %
Colour value Max. 10 APHA
Non-volatile matter Max. 0.005 %
Transmittance (210 nm) Min. 30 %
Transmittance (220 nm) Min. 60 %
Transmittance (230 nm) Min. 85 %
Transmittance (240 nm) Min. 95 %
Transmittance (250 nm) Min. 99 %