Ortofosforsyre 85% ≥85%, HiPerSolv CHROMANORM® for HPLC

Leverandør: VWR Chemicals
Danger

Synonymer: Phosphoric acid

153154DEA 560 NOK
153154D
Ortofosforsyre 85% ≥85%, HiPerSolv CHROMANORM® for HPLC
Ortofosforsyre
Formel: H₃PO₄
MW: 98 g/mol
Kokepunkt: 158
Smeltepunkt: 21
Tetthet: 1.689 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011340
CAS-nummer: 7664-38-2
EINECS: 231-633-2
FN: 1805
ADR: 8 III
Merck Indeks: 14,07344

Bestill nå

Spesifikasjon Testresultater

Assay Min. 85.0 %
Volatile acids Max. 0.0002 meq/g
Fe (Iron) Max. 10 ppm
Pb (Lead) Max. 2 ppm
Transmittance (254 nm) (0,1 mol/l) Min. 80 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR