Ammonium acetate ≥98.0%, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Warning

Synonyms: Acetic acid ammonium salt

153164REA 505 NOK
153164R
Ammonium acetate ≥98.0%, HiPerSolv CHROMANORM® for HPLC
Ammonium acetate
Formula: H₃CCOONH₄
MW: 77.08 g/mol
Melting Pt: 114
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98.0 %
pH (20°C; 5 %) 6.5 - 7.5
Fe (Iron) Max. 1 ppm
Pb (Lead) Max. 1 ppm
Transmittance (254 nm) (1 mol/l) Min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR