Potassium dihydrogen phosphate ≥99.5%, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals

Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

153184UEA 2300 NOK
153184U
Potassium dihydrogen phosphate ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Potassium dihydrogen phosphate
Formula: KH₂PO₄
MW: 136.09 g/mol
Boiling Pt: ~100 °C (1013 hPa)
Melting Pt: 253
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Order Now

Specification Test Results

Assay Min. 99.5 %
pH (20°C; 5 %) 4.3 - 4.5
Heavy metals (as Pb) Max. 10 ppm
Cl (Chloride) Max. 5 ppm
SO4 (Sulphate) Max. 30 ppm
Fe (Iron) Max. 10 ppm
Transmittance (254 nm) (0.1 mol/l) Min. 90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR