Natriumklorid ≥99.9%, HiPerSolv CHROMANORM® for HPLC

Leverandør: VWR Chemicals

Synonymer: Saline, NaCl, Sea salt, Halite

153274VEA 765 NOK
153274V
Natriumklorid ≥99.9%, HiPerSolv CHROMANORM® for HPLC
Natriumklorid
Formel: NaCl
MW: 58.44 g/mol
Kokepunkt: 1413
Smeltepunkt: 801
Tetthet: 2.16 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003477
CAS-nummer: 7647-14-5
EINECS: 231-598-3
Merck Indeks: 14,08599

Bestill nå

Spesifikasjon Testresultater

Assay (argentometric, after ignition) Min. 99.9 %
Identity Passes test
pH-value (5 %; water) 5.0 - 8.0
Transmission (206 nm; 1 mol/L; 1 cm) Min. 10 %
Transmission (254 nm; 1 mol/L; 1 cm) Min. 95 %
Br (Bromide) Max. 0.005 %
I (Iodide) Max. 0.001 %
PO4 (Phosphate) Max. 0.0005 %
SO4 (Sulphate) Max. 0.001 %
Fe (Iron) Max. 0.0001 %
K (Potassium) Max. 0.005 %
Pb (Lead) Max. 0.0002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR