Amoniakkløsning 32%, HiPerSolv CHROMANORM® for HPLC

Leverandør: VWR Chemicals
Danger

Synonymer: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

153312KEA 520 NOK
153312K
Amoniakkløsning 32%, HiPerSolv CHROMANORM® for HPLC
Amoniakkløsning
Formel: NH₃
MW: 35.05 g/mol
Kokepunkt: 32…37.7 °C (1013 hPa)
Smeltepunkt: –60…–57.5 °C
Tetthet: 0.88 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00066650
CAS-nummer: 1336-21-6
EINECS: 215-647-6
FN: 2672
ADR: 8 III

Bestill nå

Spesifikasjon Testresultater

Non-volatile residue Max. 20 ppm
Fe (Iron) Max. 0.2 ppm
Pb (Lead) Max. 0.05 ppm
Transmittance (215 nm) (1 mol/l) Min. 10 %
Transmittance (254 nm) (1 mol/l) Min. 97 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR