(±)-2-Metyltetrahydrofuran ≥99.5% stabilised, Reagent Grade

Leverandør: Honeywell Chemicals
Danger


Stabilisering: 150-400 ppm Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

155810-500ML 155810-4L 155810-2.5L
155810-500MLEA 1825 NOK
155810-500ML 155810-4L 155810-2.5L
(±)-2-Metyltetrahydrofuran ≥99.5% stabilised, Reagent Grade
(±)-2-Metyltetrahydrofuran

  • ≥95% for general laboratory use.


Beilstein Number: 102448
RTECS Number: LU2800000
UNSPSC Code: 12190000

Formel: C₅H₁₀O
MW: 86.13 g/mol
Kokepunkt: 78…80 °C (1.013 hPa)
Smeltepunkt: <–20 °C
Tetthet: 0.86 g/cm³ (20 °C)
Flash punkt: –10 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005367
CAS-nummer: 96-47-9
EINECS: 202-507-4
FN: 2536
ADR: 3 II

Bestill nå

Spesifikasjon Testresultater

Assay (GC) Min. 99.50 %
Assay BHT (GC) 150 - 400 ppm
Water (Karl Fischer) Max. 1.0 %
IR spectrum complying
Peroxides (as H₂O₂) Max. 100 ppm
Contains 150-400 ppm BHT as stabilizer
Refractive Index n20/D 1.406 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR