Tris(2-ethylhexyl)phosphate

Leverandør: Thermo Fisher Scientific
Warning

15808.18 15808.30 15808.A1
15808.18EA 375 NOK
15808.18 15808.30 15808.A1
Tris(2-ethylhexyl)phosphate
Tris(2-ethylhexyl)phosphate

CAS No.: 78-42-2 Molecular Formula: [CH3(CH2)3CH(C2H5)CH2O]3P(O) Formula Weight: 434.64 Flash Point: 110°(230°F) Physical Form: Liquid Density: 0.93 Refractive Index: 1.4410 Boiling Point: 216°/4mm MDL No.: MFCD00009491

Formel: C₂₄H₅₁O₄P
Kokepunkt: 216 °C (4 mmHg)
Tetthet: 0,93
Flash punkt: >110 °C (230 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009491
CAS-nummer: 78-42-2
EINECS: 201-116-6

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR