Tetrakloretylen, Supelco®

Leverandør: Merck
Warning

Synonymer: Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene

M-Clarity™ quality level = MQ100

15889-10ML-F
15889-10ML-FEA 2550 NOK
15889-10ML-F
Tetrakloretylen, Supelco®
Tetrakloretylen
Formel: C₂Cl₄
MW: 165.83 g/mol
Kokepunkt: 121.1
Smeltepunkt: –22 °C
Tetthet: 1.63 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000834
CAS-nummer: 127-18-4
EINECS: 204-825-9
FN: 1897
ADR: 6.1 III
Merck Indeks: 12,09332
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=509ab4de043c3b0d929a6b7531f507505437ea13&vid=71e4bead215afb2af05e4712d1aa0e447182f388&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR