EDTA dinatrium salt 0.01 mol/l (0.02 N) in aqueous solution, AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals

Synonymer: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

160226GEA 1300 NOK
160226G
EDTA dinatrium salt 0.01 mol/l (0.02 N) in aqueous solution, AVS TITRINORM volumetric solution
EDTA dinatrium salt
Formel: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00070672
CAS-nummer: 139-33-3
EINECS: 205-358-3
Merck Indeks: 12,03556

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.00998 - 0.01002 mol/l
NIST traceable Confirmed

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR