Brilliant cresyl blue, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: CI 51010, 1,3-Diamino-7-(diethylamino)-3-methylphenoxazin-5-ium tetrachlorozincate (2:1), Brilliant cresyl blue zinc chloride double salt, Brilliant Blue C, BCB

16030-10G 16030-50G
16030-10GEA 867 NOK
16030-10G 16030-50G
Brilliant cresyl blue, Sigma-Aldrich®
Brilliant cresyl blue

'Brilliant Cresyl blue',for microscopy (Vit.), mixture of toluidine blue and waterblue ('Cresyl blue')

Formel: C₃₄H₄₂Cl₄N₈O₂Zn
MW: 801.96 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00148901
CAS-nummer: 81029-05-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR