Fenol rødt ACS

Leverandør: Thermo Fisher Scientific
Warning

16294.06 16294.14 16294.22
16294.06EA 560 NOK
16294.06 16294.14 16294.22
Fenol rødt ACS
Fenol rødt

CAS No.: 143-74-8 Molecular Formula: C19H14O5S Formula Weight: 354.38 Physical Form: Crystalline Odour: Odourless Melting Point: 300° Beilstein Reference: 326470 Merck Reference: 14,7247 MDL No.: MFCD00003552


pH indicator

Formel: C₁₉H₁₄O₅S
Smeltepunkt: >300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003552
CAS-nummer: 143-74-8
EINECS: 205-609-7
Merck Indeks: 14,7247

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR