Electrolyte solution, non-aqueous, Supelco®
Leverandør: Merck
16398-250ML
16398-250MLEA 680 NOK
16398-250ML
Electrolyte solution, non-aqueous, Supelco®
Elektrolytter Løsninger, elektroder
Lithium chloride solution is used for the determination of trace alkaline earth metals in brines, via chelation ion chromatography, on iminodiacetic acid bonded silica column.
It may also be used along with N,N-dimethylacetamide (DMAC) may be used in size exclusion chromatography of cellulose.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cd3cdd86af890df959ecdb7b9d36df14f4dc400&vid=7162ace03eb84896d705ca2d2024d76b64ebeca6&prid=web&clid=SAPDAM