7-Amino-4-methylcoumarin, Sigma-Aldrich®

Leverandør: Merck Millipore Calbiochem

164545-10
164545-10EA 2000 NOK
164545-10
7-Amino-4-methylcoumarin, Sigma-Aldrich®
7-Amino-4-methylcoumarin
Formel: C₁₀H₉NO₂
Kokepunkt: 352 °C (1013 hPa)
Smeltepunkt: 226 °C
Tetthet: 1,377 g/cm³ (20 °C)
MDL nummer: MFCD00006868
CAS-nummer: 26093-31-2
EINECS: 247-454-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR