Tannin ≥96.0% (dry basis) FCC, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials

1674-21EA 32475 NOK
1674-21
Tannin ≥96.0% (dry basis) FCC, Macron Fine Chemicals™
Tannic acid

  • Food GMP manufactured product

Formel: C₇₆H₅₂O₄₆
Smeltepunkt: 218…219 °C
Tetthet: >1 g/cm³ (20 °C)
Flash punkt: 199 °C
MDL nummer: MFCD00066397
CAS-nummer: 1401-55-4
EINECS: 215-753-2
Merck Indeks: 12,09221

Bestill nå

Spesifikasjon Testresultater

Food GMP Manufactured Product ≥96.0%
Meets F.C.C. Requirements Passes Test
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Food Chemical Passes Test
Assay () (dried basis) (FCC) ≥96.0 %
Appearance (fluffy, light tan to brown powder) Passes Test
Solubility Passes Test
Gums or Dextrin Passes Test
Resinous Substances Passes Test
Identification A Passes Test
Identification B Passes Test
Loss on Drying at 105°C ≤7.0 %
Residue on Ignition ≤1.0 %
Lead (Pb) ≤2 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR