Supelco® Organic Reference Standard, Diuron
Leverandør: Merck
16902-50MG
16902-50MGEA 825 NOK
16902-50MG
Supelco® Organic Reference Standard, Diuron
Standarder Organiske standarder
  • GHS signal: GHS08, GHS07, GHS09
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
NB: P202 - P260 - P264 - P273 - P301 + P312 - P308 + P313
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7dcf8d4afc77d42fc76ebefbed11ce035fd1c055&vid=7dcf8d4afc77d42fc76ebefbed11ce035fd1c055&prid=web&clid=SAPDAM
Bestill nå
SPESIFIKASJONER
Synonym(s) 3-(3,4-Dichlorophenyl)-1,1-dimethylurea
Environmentally Preferable
M-Clarity quality level MQ300
CAS-nr 330-54-1
MDL No. MFCD00018136
Molekylvekt 233,09
Hardhetsklasse beskrivelse H302 - H351 - H373 - H410
Flammepunkt Not applicable
Fraktforhold STD
Lagringsforhold 15...25 °C