Gel filtration media, Sephadex™ G-15

Leverandør: Cytiva

17-0020-01
17-0020-01EA 3825 NOK
17-0020-01
Gel filtration media, Sephadex™ G-15
Proteinrense Protein Purification Resins

Sephadex™ G-15 is a well established gel filtration medium for desalting and buffer exchange of peptides and small biomolecules >1500 molecular weight.


  • Quickly desalts, removes contaminants and transfers to a new buffer in a single step
  • Suitable for buffer exchange and cleanup of biological samples, for example peptides, small proteins, and oligosaccharides larger than 1500 molecular weight
  • Efficient removal of contaminants such as salts, dyes, and radioactive labels


Sephadex™ is a gel filtration medium prepared by crosslinking dextran with epichlorohydrin. Different types of Sephadex™ differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. Sephadex™ G-15 is one of six different G-types ranging from G-10 for small molecules to G-100 for very large biomolecules.

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Matrix Cross-linked dextran
fraksjoneringsspekter <1,5 x 10³ dextrans, <1,5 x 10³ globular proteins
pH område 2 - 13
Lagringstemp 4 - 30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR