Sepharose™ CL-4B Phenyl Hydrophobic Interaction Chromatography Media

Leverandør: Cytiva

17-0810-01
17-0810-01EA 8450 NOK
17-0810-01
Sepharose™ CL-4B Phenyl Hydrophobic Interaction Chromatography Media
Proteinrense Protein Purification Resins

Octyl-Sepharose™ CL-4B is a well-proven medium for hydrophobic interaction chromatography (HIC). Octyl immobilised on 4% cross-linked agarose.


  • Phenyl immobilized on 4% cross-linked agarose
  • Aromatic ligand
  • Stable in all commonly used aqueous buffers and additives such as 8 M urea and 6 M guanidine hydrochloride
  • Negligible volume variation due to changes in pH or ionic strength


Octyl Sepharose® CL-4B and Phenyl Sepharose® CL-4B are covalently coupled to a cross-linked 4% agarose matrix by ether linkage, resulting in hydrophobic gels with minimal leakage and no ionic properties.

Bestill nå

SPESIFIKASJONER

Ligand Phenyl
Environmentally Preferable
Liganddensitet 40 μmole/ml drained medium
Matrix Cross-linked agarose, 4%, spherical
Gjennomstrømningshastighet 150 cm/h
pH område 3 - 12
Bindingskapasitet HSA (Mr 68 000) 15-20 mg/ml drained resin, ß-lactoglobulin (Mr 18 000) 3-5 mg/ml drained resin
Lagringstemp 4 - 30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR