Vinylferrocene

Leverandør: Thermo Fisher Scientific

17450.03 17450.06
17450.03EA 1625 NOK
17450.03 17450.06
Vinylferrocene
Vinylferrocene

CAS No.: 1271-51-8 Molecular Formula: C12H12Fe Formula Weight: 212.07 Flash Point: 62°(144°F) Physical Form: Powder Melting Point: 52-55° Boiling Point: 80-85°/0.7mm MDL No.: MFCD00001434

Formel: C₁₂H₁₂Fe
Kokepunkt: 80…85 °C (0,7 mmHg)
Smeltepunkt: 52…55 °C
Flash punkt: 62 °C (144 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001434
CAS-nummer: 1271-51-8
EINECS: 215-041-1

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR